Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hesperidin Methyl Chalcone, MP Biomedicals

Catalog No. ICN215520 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN215520 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN215520 Supplier MP Biomedicals, Inc Supplier No. 0521552010

Chemical Identifiers

CAS 24292-52-2
Molecular Formula C29H36O15
Molecular Weight (g/mol) 624.59
InChI Key FDHNLHLOJLLXDH-JIYHLSBYSA-N
Synonym hesperidin methylchalcone, hesperidin methyl chalcone, unii-4t2gva922x, e-1-4-6-o-6-deoxy-alpha-l-mannopyranosyl-beta-d-glucopyranosyl oxy-2-hydroxy-6-methoxyphenyl-3-3-hydroxy-4-methoxyphenyl-2-propen-1-one, 1-4-6-o-6-deoxy-alpha-l-mannopyranosyl-beta-d-glucopyranosyl oxy-2-hydroxy-6-methoxyphenyl-3-3-hydroxy-4-methoxyphenyl prop-2-en-1-one, 2-propen-1-one, 1-4-6-o-6-deoxy-alpha-l-mannopyranosyl-beta-d-glucopyranosyl oxy-2-hydroxy-6-methoxyphenyl-3-3-hydroxy-4-methoxyphenyl-, 2e, chalcone, 2',3,4'-trihydroxy-4,6'-dimethoxy-, 4'-6-o-6-deoxy-alpha-l-mannopyranosyl-beta-d-glucopyranoside, e-1-2-methoxy-4-2s,3r,4s,5s,6r-6-2r,3r,4r,5r,6s-6-methyl-3,4,5-tris oxidanyl oxan-2-yl oxymethyl-3,4,5-tris oxidanyl oxan-2-yl oxy-6-oxidanyl-phenyl-3-4-methoxy-3-oxidanyl-phenyl prop-2-en-1-one, e-1-4-6-o-6-deoxy-, a-l-mannopyranosyl-, a-d-glucopyranosyl oxy-2-hydroxy-6-methoxyphenyl-3-3-hydroxy-4-methoxyphenyl-2-propen-1-one, e-3-3-hydroxy-4-methoxyphenyl-1-2-hydroxy-6-methoxy-4-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-2r,3r,4r,5r,6s-3,4,5-trihydroxy-6-methyloxan-2-yl oxymethyl oxan-2-yl oxyphenyl prop-2-en-1-one
PubChem CID 6436550

Specifications

Quantity 10 g
Chemical Name or Material Hesperidin Methyl Chalcone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.