accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 24300-91-2

CH3CH3CH3H3CCH3CH3

CAS RN 24300-91-2

IUPAC Name: 2,7-di-tert-butylpyrene
Synonyms: 2,7-di-tert-butylpyrene
Molecular Weight (g/mol): 314.47
Molecular Formula: C24H26
InChi Key: SKZSSCAGJZEXEA-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC2=CC=C3C=C(C=C4C=CC(=C1)C2=C34)C(C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)