Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,7-Di-tert-butylpyrene 98.0+%, TCI America™
SDP

Catalog No. D41855G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41855G 5 g
D41851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D41855G Supplier TCI America Supplier No. D41855G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 24300-91-2
Molecular Formula C24H26
Molecular Weight (g/mol) 314.47
MDL Number MFCD01013641
InChI Key SKZSSCAGJZEXEA-UHFFFAOYSA-N
PubChem CID 185563
IUPAC Name 2,7-di-tert-butylpyrene
SMILES CC(C)(C)C1=CC2=CC=C3C=C(C=C4C=CC(=C1)C2=C34)C(C)(C)C

Specifications

Melting Point 209°C
Color White-Yellow
Quantity 5 g
Formula Weight 314.47
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Di-tert-butylpyrene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.