accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONF

CAS RN 243128-37-2

IUPAC Name: 4-fluoro-3-methoxybenzonitrile
Synonyms: 4-fluoro-3-methoxybenzonitrile
Molecular Weight (g/mol): 151.14
Molecular Formula: C8H6FNO
InChi Key: FOWHAPVFVBXMBK-UHFFFAOYSA-N
SMILES: COC1=C(F)C=CC(=C1)C#N
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results