Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-3-methoxybenzonitrile, TCI America™
SDP

Catalog No. F04081G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F04081G Supplier TCI America Supplier No. F04081G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 243128-37-2
Molecular Formula C8H6FNO
Molecular Weight (g/mol) 151.14
MDL Number MFCD01569371
InChI Key FOWHAPVFVBXMBK-UHFFFAOYSA-N
Synonym 3-methoxy-4-fluorobenzonitrile, 4-fluoro-3-methoxy benzonitrile, benzonitrile, 4-fluoro-3-methoxy, 4-fluoro-3-methoxy-benzonitrile, 4-fluoro-3-methoxybenzenecarbonitrile, pubchem3501, acmc-209gaq, 5-cyano-2-fluoroanisole, intermediates-zcf02653, ksc494k3r
PubChem CID 2737365
IUPAC Name 4-fluoro-3-methoxybenzonitrile
SMILES COC1=C(F)C=CC(=C1)C#N

Specifications

Melting Point 109°C
Quantity 1 g
UN Number 3439
Formula Weight 151.14
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-3-methoxybenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.