accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OHO

CAS RN 24323-21-5

IUPAC Name: 2-methylnonanoic acid
Synonyms: 2-methyl nonaoic acid
Molecular Weight (g/mol): 172.27
Molecular Formula: C10H20O2
InChi Key: KUSYIGBGHPOWEL-UHFFFAOYNA-N
SMILES: CCCCCCCC(C)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results