Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylnonanoic acid, 98%, Thermo Scientific™

Catalog No. AAH3043006
Change view
Click to view available options
Quantity:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3043006 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3043006 Supplier Thermo Scientific Chemicals Supplier No. H3043006

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 24323-21-5
Boiling Point 265°C
Odor Unpleasant
Molecular Formula C10H20O2
Quantity 5g
MDL Number MFCD00154319
UN Number UN3265
Synonym 2-methyl nonaoic acid, 2-methylnonan-1-oic acid, nonanoic acid, 2-methyl, 2-methyl-nonanoic acid
InChI Key KUSYIGBGHPOWEL-UHFFFAOYSA-N
SMILES CCCCCCCC(C)C(=O)O
IUPAC Name 2-methylnonanoic acid
Molecular Weight (g/mol) 172.268
PubChem CID 91166
Formula Weight 172.27
Percent Purity 98%
Chemical Name or Material 2-Methylnonanoic acid
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 246-162-8
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.