accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOH3COHOHOH

CAS RN 24332-98-7

IUPAC Name: 2-methoxy-6-methyloxane-3,4,5-triol
Synonyms: (-)-fucose 6-deoxy-L-β-galactose rhodeose isodulcit mannomethylose, L- L-(-)-fucose l-(+)-rhamnose monohydrate uridine 5-pyrophosphate L-galactose, 6-deoxy- L-fuc
Molecular Weight (g/mol): 178.18
Molecular Formula: C7H14O5
InChi Key: OHWCAVRRXKJCRB-UHFFFAOYNA-N
SMILES: COC1OC(C)C(O)C(O)C1O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results