CAS RN 24345-16-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Apamin, Bee Venom
Polypeptide neurotoxin known to pass through the blood-brain barrier.