Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Apamin, Bee Venom

Catalog No. 178270500UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-827-0500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-827-0500UG Supplier MilliporeSigma™ Supplier No. 178270500UG
Add to Cart
Add to Cart

Polypeptide neurotoxin known to pass through the blood-brain barrier.

Peptide Sequence: H-Cys1-Asn-Cys3-Lys-Ala-Pro-Glu-Thr-Ala-Leu-Cys11-Ala-Arg-Arg-Cys15-Gln-Gln-His-NH2 (disulfide bonds: 1 → 11; 3 → 15)
TRUSTED_SUSTAINABILITY

Specifications

Color White with Dark Tan Cast
Description 98% by HPLC
Target Proteins AHP-type Ca2+-activated K+ channel
CAS 24345-16-2
Molecular Formula C79H131N31O24S4
Formula Weight 2027.3g/mol
Solubility 5% Acetic Acid
Content And Storage Hygroscopic
Form Solid
Quantity 500 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.