accessibility menu, dialog, popup

UserName

Viewing profile as user:
FHOFFF

CAS RN 243459-91-8

IUPAC Name: 5-fluoro-2-(trifluoromethyl)phenol
Synonyms: 5-fluoro-2-(trifluoromethyl)phenol
Molecular Weight (g/mol): 180.1
Molecular Formula: C7H4F4O
InChi Key: QWLZSSYHAJSEHU-UHFFFAOYSA-N
SMILES: OC1=C(C=CC(F)=C1)C(F)(F)F
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results