accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 24363-37-9

IUPAC Name: barium(2+) bis(propan-2-olate)
Synonyms: barium(2+) bis(propan-2-olate)
Molecular Weight (g/mol): 255.5
Molecular Formula: C6H14BaO2
InChi Key: CPUJSIVIXCTVEI-UHFFFAOYSA-N
SMILES: [Ba++].CC(C)[O-].CC(C)[O-]
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)