Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium isopropoxide, 99.5% (metals basis), Thermo Scientific™

Catalog No. AA4127114
Change view
Click to view available options
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AA4127114 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4127114 Supplier Thermo Scientific Chemicals Supplier No. 4127114

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 24363-37-9
Molecular Formula C6H14BaO2
Linear Formula Ba[OCH(CH3)2]2
Quantity 25g
MDL Number MFCD00050486
UN Number UN3205
Sensitivity Moisture sensitive
Synonym barium isopropoxide, barium 2+ ; propan-2-olate, barium diisopropoxide, acmc-20alr0, barium ii isopropoxide, barium 2+ bis propan-2-olate, barium 2+ ion bis propan-2-olate
Solubility Information Soluble in alcohols,ketones and esters. Reacts with water.
InChI Key CPUJSIVIXCTVEI-UHFFFAOYSA-N
SMILES CC(C)[O-].CC(C)[O-].[Ba+2]
IUPAC Name barium(2+);propan-2-olate
Molecular Weight (g/mol) 255.503
PubChem CID 6101105
Formula Weight 255.52
Percent Purity 99.5%
Assay (metals basis)
Chemical Name or Material Barium isopropoxide
Show More Show Less
Hazard Category H228-H252-H302+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318-H228-H252-H302-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
Self-heating in large quantities
may catch fire.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P210-P235+P410-P240-P241-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.2; Packing Group: III; Proper Shipping Name: ALKALINE EARTH METAL ALCOHOLATES, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.