accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 243669-48-9

IUPAC Name: ethyl 5'-amino-[2,3'-bithiophene]-4'-carboxylate
Synonyms: ethyl 5'-amino-[2,3'-bithiophene]-4'-carboxylate
Molecular Weight (g/mol): 253.33
Molecular Formula: C11H11NO2S2
InChi Key: AKFDBWIUWIWHRK-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(N)SC=C1C1=CC=CS1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results