Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-amino-4-(2-thienyl)thiophene-3-carboxylate, 97%, Thermo Scientific™

Catalog No. AAH5877706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5877706 5 g
AAH5877703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5877706 Supplier Thermo Scientific Chemicals Supplier No. H5877706

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 243669-48-9
Molecular Formula C11H11NO2S2
Molecular Weight (g/mol) 253.33
MDL Number MFCD00115120
InChI Key AKFDBWIUWIWHRK-UHFFFAOYSA-N
Synonym ethyl 2-amino-4-2-thienyl thiophene-3-carboxylate, ethyl 5'-amino-2,3'-bithiophene-4'-carboxylate, ethyl 2-amino-4-thiophen-2-yl thiophene-3-carboxylate, 5'-amino-2,3' bithiophenyl-4'-carboxylic acid ethyl ester, ethyl 5'-amino 2,3'-bithiophene-4'-carboxylate, ethyl 2-azanyl-4-thiophen-2-yl-thiophene-3-carboxylate, 2,3'-bithiophene-4'-carboxylicacid, 5'-amino-, ethyl ester, 2-amino-4-thiophen-2-yl-3-thiophenecarboxylic acid ethyl ester, maybridge1_003774
PubChem CID 718227
SMILES CCOC(=O)C1=C(N)SC=C1C1=CC=CS1

Specifications

Melting Point 85°C to 87°C
Quantity 5 g
IUPAC Name ethyl 5'-amino-[2,3'-bithiophene]-4'-carboxylate
Formula Weight 253.33
Percent Purity 97%
Chemical Name or Material Ethyl 2-amino-4-(2-thienyl)thiophene-3-carboxylate
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.