accessibility menu, dialog, popup

UserName

Viewing profile as user:
OSNO

CAS RN 24372-49-4

IUPAC Name: 5-(morpholin-4-yl)thiophene-2-carbaldehyde
Synonyms: 5-(morpholin-4-yl)thiophene-2-carbaldehyde
Molecular Weight (g/mol): 197.25
Molecular Formula: C9H11NO2S
InChi Key: MWCGENRKAMIZLZ-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(S1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results