accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3(S)OOH(S)OH(R)HO(S)HO

CAS RN 2438-80-4

IUPAC Name: (3S,4R,5S,6S)-6-methyloxane-2,3,4,5-tetrol
Synonyms: fucose L-fucopyranose 6-deoxy-L-galactose L-fucose
Molecular Weight (g/mol): 164.16
Molecular Formula: C6H12O5
InChi Key: SHZGCJCMOBCMKK-DHVFOXMCSA-N
SMILES: C[C@@H]1OC(O)[C@@H](O)[C@H](O)[C@@H]1O
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (3)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)