accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 24386-93-4

IUPAC Name: 2-{4-amino-5-iodo-7H-pyrrolo[2,3-d]pyrimidin-7-yl}-5-(hydroxymethyl)oxolane-3,4-diol
Synonyms: 2-{4-amino-5-iodopyrrolo[2,3-d]pyrimidin-7-yl}-5-(hydroxymethyl)oxolane-3,4-diol
Molecular Weight (g/mol): 392.15
Molecular Formula: C11H13IN4O4
InChi Key: WHSIXKUPQCKWBY-UHFFFAOYNA-N
SMILES: NC1=C2C(I)=CN(C3OC(CO)C(O)C3O)C2=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results