accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 24387-36-8

IUPAC Name: 3,5-diphenyl-2-{4-[(1E)-2-phenylethenyl]phenyl}-3H-1,2λ⁵,3,4-tetrazol-2-ylium chloride
Synonyms: 3,5-diphenyl-2-{4-[(1E)-2-phenylethenyl]phenyl}-1,2λ⁵,3,4-tetrazol-2-ylium chloride
Molecular Weight (g/mol): 436.94
Molecular Formula: C27H21ClN4
InChi Key: PAKWFHYWNFCRCK-CMBBICFISA-M
SMILES: [Cl-].C(=C/C1=CC=C(C=C1)[N+]1=NC(=NN1C1=CC=CC=C1)C1=CC=CC=C1)\C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results