Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Diphenyl-3-(4-styrylphenyl)tetrazolium Chloride 96.0+%, TCI America™
SDP

Catalog No. D09151G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D09151G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D09151G Supplier TCI America Supplier No. D09151G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 24387-36-8
Molecular Formula C27H21ClN4
Molecular Weight (g/mol) 436.94
MDL Number MFCD00050315
InChI Key PAKWFHYWNFCRCK-CMBBICFISA-M
PubChem CID 9932775
IUPAC Name 3,5-diphenyl-2-{4-[(1E)-2-phenylethenyl]phenyl}-3H-1,2λ⁵,3,4-tetrazol-2-ylium chloride
SMILES [Cl-].C(=C/C1=CC=C(C=C1)[N+]1=NC(=NN1C1=CC=CC=C1)C1=CC=CC=C1)\C1=CC=CC=C1

Specifications

Melting Point 223°C
Color Yellow
Quantity 1 g
Formula Weight 436.94
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Diphenyl-3-(4-styrylphenyl)tetrazolium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.