accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONH2O

CAS RN 2439-77-2

IUPAC Name: 2-methoxybenzamide
Synonyms: O-methoxybenzamide
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: MNWSGMTUGXNYHJ-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results