accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOCH3(E/Z)

CAS RN 24393-56-4

IUPAC Name: ethyl 3-(4-methoxyphenyl)prop-2-enoate
Synonyms: ethyl 3-(4-methoxyphenyl)prop-2-enoate
Molecular Weight (g/mol): 206.24
Molecular Formula: C12H14O3
InChi Key: DHNGCHLFKUPGPX-UHFFFAOYSA-N
SMILES: CCOC(=O)C=CC1=CC=C(OC)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results