Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-Methoxycinnamate 98.0+%, TCI America™
SDP

Catalog No. M12045G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12045G 5 g
M120425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M12045G Supplier TCI America Supplier No. M12045G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 24393-56-4
Molecular Formula C12H14O3
Molecular Weight (g/mol) 206.241
MDL Number MFCD00026906
InChI Key DHNGCHLFKUPGPX-RMKNXTFCSA-N
Synonym 4-Methoxycinnamic Acid Ethyl Ester, Ethyl 3-(4-Methoxyphenyl)acrylate, 3-(4-Methoxyphenyl)acrylic Acid Ethyl Ester
PubChem CID 5281783
ChEBI CHEBI:29015
IUPAC Name ethyl (E)-3-(4-methoxyphenyl)prop-2-enoate
SMILES CCOC(=O)C=CC1=CC=C(C=C1)OC

Specifications

Melting Point 49°C
Color White
Boiling Point 187°C
Quantity 5 g
Formula Weight 206.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 4-Methoxycinnamate
EINECSNumber (3)-1706
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.