CAS RN 243966-09-8
IUPAC Name:
2-(carboxyformamido)-4H,5H,6H,7H-thieno[2,3-c]pyridine-3-carboxylic acid hydrochloride
Synonyms:
2-(carboxyformamido)-4H,5H,6H,7H-thieno[2,3-c]pyridine-3-carboxylic acid hydrochloride
Molecular Weight (g/mol):
306.72
Molecular Formula:
C10H11ClN2O5S
InChi Key:
LQGCAMWQDSYOAY-UHFFFAOYSA-N
SMILES:
Cl.OC(=O)C(=O)NC1=C(C(O)=O)C2=C(CNCC2)S1
1
–
1
of
1
results
1
TCS 401, Tocris Bioscience™
CAS: 243966-09-8 Molecular Formula: C10H11ClN2O5S Molecular Weight (g/mol): 306.717 InChI Key: LQGCAMWQDSYOAY-UHFFFAOYSA-N Synonym: 2-carboxycarbonyl amino-4,5,6,7-tetrahydrothieno 2,3-c pyridine-3-carboxylic acid hydrochloride,2-carboxyformamido-4h,5h,6h,7h-thieno 2,3-c pyridine-3-carboxylic acid hydrochloride PubChem CID: 56972185 IUPAC Name: 2-(oxaloamino)-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxylic acid;hydrochloride SMILES: C1CNCC2=C1C(=C(S2)NC(=O)C(=O)O)C(=O)O.Cl