Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCS 401, R&D Systems™
SDP

Catalog No. 275410
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-541-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 27-541-0 Supplier R&D Systems Supplier No. 2754/10

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective inhibitor of PTP1B

Selective inhibitor of protein-tyrosine phosphatase 1B (PTP1B) (Ki values are 0.29, 59, 560, 1100, > 2000, > 2000 and > 2000 μM for PTP1B, CD45 D1D2, PTPβ, PTPε D1, SHP-1, PTPα D1 and LAR D1D2 respectively).

Chemical Identifiers

CAS 243966-09-8
Molecular Formula C10H11ClN2O5S
Molecular Weight (g/mol) 306.717
InChI Key LQGCAMWQDSYOAY-UHFFFAOYSA-N
Synonym 2-carboxycarbonyl amino-4,5,6,7-tetrahydrothieno 2,3-c pyridine-3-carboxylic acid hydrochloride, 2-carboxyformamido-4h,5h,6h,7h-thieno 2,3-c pyridine-3-carboxylic acid hydrochloride
PubChem CID 56972185
IUPAC Name 2-(oxaloamino)-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxylic acid;hydrochloride
SMILES C1CNCC2=C1C(=C(S2)NC(=O)C(=O)O)C(=O)O.Cl

Specifications

Quantity 10 mg
Formula Weight 306.72
Percent Purity >98%
Product Line Tocris
Chemical Name or Material TCS 401
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.