accessibility menu, dialog, popup

UserName

Viewing profile as user:
Cl

CAS RN 2441-97-6

IUPAC Name: 3-chlorocyclohex-1-ene
Synonyms: 3-chlorocyclohex-1-ene
Molecular Weight (g/mol): 116.59
Molecular Formula: C6H9Cl
InChi Key: LNGQLHZIYFQUIR-UHFFFAOYNA-N
SMILES: ClC1CCCC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results