Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chlorocyclohexene 95.0+%, TCI America™
SDP

Catalog No. C15635ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15635ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C15635ML Supplier TCI America Supplier No. C15635ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2441-97-6
Molecular Formula C6H9Cl
Molecular Weight (g/mol) 116.588
MDL Number MFCD00013776
InChI Key LNGQLHZIYFQUIR-UHFFFAOYSA-N
Synonym 1,2,3,4-Tetrahydrochlorobenzene
PubChem CID 17120
IUPAC Name 3-chlorocyclohexene
SMILES C1CC=CC(C1)Cl

Specifications

Color Yellow
Boiling Point 146°C
Quantity 5 mL
UN Number 1993
Formula Weight 116.59
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chlorocyclohexene
RTECSNumber GW4605000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.