accessibility menu, dialog, popup

UserName

Viewing profile as user:
BFFFFH3CNNCH3

CAS RN 244193-56-4

IUPAC Name: 1-decyl-3-methyl-1H-imidazol-3-ium; tetrafluoroboranuide
Synonyms: 1-decyl-3-methylimidazolium tetrafluoroborate
Molecular Weight (g/mol): 310.19
Molecular Formula: C14H27BF4N2
InChi Key: QGUMDWFYJYXDTH-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.CCCCCCCCCCN1C=C[N+](C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results