Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S,S)-2,3-Bis(Tert-Butylmethylphosphino)Quinoxaline

Catalog No. 694215100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-421-5100MG 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 69-421-5100MG Supplier Sigma Aldrich Supplier No. 694215100MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S,S)-2,3-Bis(Tert-Butylmethylphosphino)Quinoxaline

Specifications

CAS 244193-56-4
Molecular Formula C14H27BF4N2
Linear Formula C14H27BF4N2
Refractive Index n20/D 1.438-1.450
MDL Number MFCD09038853
Quantity 50 g
Molecular Weight (g/mol) 310.18 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.