CAS RN 2445-76-3
IUPAC Name:
hexyl propanoate
Synonyms:
hexyl propionate
Molecular Weight (g/mol):
158.24
Molecular Formula:
C9H18O2
InChi Key:
GOKKOFHHJFGZHW-UHFFFAOYSA-N
SMILES:
CCCCCCOC(=O)CC
1
–
1
of
1
results
1
Hexyl Propionate 98.0+%, TCI America™
CAS: 2445-76-3 Molecular Formula: C9H18O2 Molecular Weight (g/mol): 158.241 MDL Number: MFCD00085197 InChI Key: GOKKOFHHJFGZHW-UHFFFAOYSA-N Synonym: Propionic Acid Hexyl Ester PubChem CID: 88454 ChEBI: CHEBI:87549 IUPAC Name: hexyl propanoate SMILES: CCCCCCOC(=O)CC