Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexyl Propionate 98.0+%, TCI America™
SDP

Catalog No. H1330500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1330500ML 500 mL
H133025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H1330500ML Supplier TCI America Supplier No. H1330500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2445-76-3
Molecular Formula C9H18O2
Molecular Weight (g/mol) 158.241
MDL Number MFCD00085197
InChI Key GOKKOFHHJFGZHW-UHFFFAOYSA-N
Synonym Propionic Acid Hexyl Ester
PubChem CID 88454
ChEBI CHEBI:87549
IUPAC Name hexyl propanoate
SMILES CCCCCCOC(=O)CC

Specifications

Color Colorless
Boiling Point 186°C
Quantity 500 mL
Formula Weight 158.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexyl Propionate
EINECSNumber (2)-0774
RTECSNumber UA2462200
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.