accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2449-35-6

IUPAC Name: 4-(4-carboxybenzenesulfonyl)benzoic acid
Synonyms: 4-(4-carboxybenzenesulfonyl)benzoic acid
Molecular Weight (g/mol): 306.29
Molecular Formula: C14H10O6S
InChi Key: SQJQLYOMPSJVQS-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)S(=O)(=O)C1=CC=C(C=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results