accessibility menu, dialog, popup

UserName

Viewing profile as user:
NN

CAS RN 2451-91-4

IUPAC Name: dibenzyl(cyano)amine
Synonyms: dibenzyl(cyano)amine
Molecular Weight (g/mol): 222.29
Molecular Formula: C15H14N2
InChi Key: GJAHYSLBRZODMY-UHFFFAOYSA-N
SMILES: N#CN(CC1=CC=CC=C1)CC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results