Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibenzylcyanamide 98.0+%, TCI America™
SDP

Catalog No. D33205G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D33205G Supplier TCI America Supplier No. D33205G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2451-91-4
Molecular Formula C15H14N2
Molecular Weight (g/mol) 222.291
InChI Key GJAHYSLBRZODMY-UHFFFAOYSA-N
Synonym N-Cyanodibenzylamine
PubChem CID 222027
IUPAC Name dibenzylcyanamide
SMILES C1=CC=C(C=C1)CN(CC2=CC=CC=C2)C#N

Specifications

Melting Point 54°C
Color White-Yellow
Boiling Point 148°C
Quantity 5 g
Formula Weight 222.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dibenzylcyanamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.