accessibility menu, dialog, popup

UserName

Viewing profile as user:
Cl

CAS RN 2453-46-5

IUPAC Name: chlorocycloheptane
Synonyms: chlorocycloheptane
Molecular Weight (g/mol): 132.63
Molecular Formula: C7H13Cl
InChi Key: KMJSGLZXFNSANB-UHFFFAOYSA-N
SMILES: ClC1CCCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results