Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorocycloheptane 98.0+%, TCI America™
SDP

Catalog No. C068110ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C068110ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C068110ML Supplier TCI America Supplier No. C068110ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2453-46-5
Molecular Formula C7H13Cl
Molecular Weight (g/mol) 132.63
MDL Number MFCD00046501
InChI Key KMJSGLZXFNSANB-UHFFFAOYSA-N
Synonym Cycloheptyl Chloride
PubChem CID 75562
IUPAC Name chlorocycloheptane
SMILES ClC1CCCCCC1

Specifications

Color Yellow
Boiling Point 175°C
Quantity 10 mL
UN Number 1993
Formula Weight 132.63
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Chlorocycloheptane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.