accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOHO

CAS RN 24539-28-4

IUPAC Name: 8-(methoxycarbonyl)cubane-1-carboxylic acid
Synonyms: 8-(methoxycarbonyl)cubane-1-carboxylic acid
Molecular Weight (g/mol): 206.2
Molecular Formula: C11H10O4
InChi Key: ARRJEFLYRAVFJA-UHFFFAOYSA-N
SMILES: COC(=O)C12C3C4C1C1C2C3C41C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results