accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 24539-57-9

H3COOHOO

CAS RN 24539-57-9

IUPAC Name: 2-[(hexyloxy)carbonyl]benzoic acid
Synonyms: 2-[(hexyloxy)carbonyl]benzoic acid
Molecular Weight (g/mol): 250.29
Molecular Formula: C14H18O4
InChi Key: XOSNGXNHDRYFEF-UHFFFAOYSA-N
SMILES: CCCCCCOC(=O)C1=CC=CC=C1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results