Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monohexyl Phthalate 98.0+%, TCI America™
SDP

Catalog No. P13135G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P13135G Supplier TCI America Supplier No. P13135G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 24539-57-9
Molecular Formula C14H18O4
Molecular Weight (g/mol) 250.294
MDL Number MFCD00517703
InChI Key XOSNGXNHDRYFEF-UHFFFAOYSA-N
Synonym Phthalic Acid Monohexyl Ester
PubChem CID 90532
IUPAC Name 2-hexoxycarbonylbenzoic acid
SMILES CCCCCCOC(=O)C1=CC=CC=C1C(=O)O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 250.29
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Monohexyl Phthalate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.