accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OOOOCH3CH3H3C

CAS RN 245652-81-7

IUPAC Name: 1,2-bis(3,3,5-trimethylcyclohexyl) benzene-1,2-dicarboxylate
Synonyms: 1,2-bis(3,3,5-trimethylcyclohexyl) phthalate
Molecular Weight (g/mol): 414.59
Molecular Formula: C26H38O4
InChi Key: ATHBXDPWCKSOLE-UHFFFAOYNA-N
SMILES: CC1CC(CC(C)(C)C1)OC(=O)C1=CC=CC=C1C(=O)OC1CC(C)CC(C)(C)C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results