Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(cis-3,3,5-trimethylcyclohexyl) Phthalate 98.0+%, TCI America™
SDP

Catalog No. P09465G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P09465G 5 g
P094625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P09465G Supplier TCI America Supplier No. P09465G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 245652-81-7
Molecular Formula C26H38O4
Molecular Weight (g/mol) 414.586
MDL Number MFCD00070480
InChI Key ATHBXDPWCKSOLE-LHQJTOJGSA-N
Synonym Phthalic Acid Bis(cis-3,3,5-trimethylcyclohexyl) Ester
PubChem CID 53661088
IUPAC Name 2-O-[(1R,5R)-3,3,5-trimethylcyclohexyl] 1-O-(3,3,5-trimethylcyclohexyl) benzene-1,2-dicarboxylate
SMILES CC1CC(CC(C1)(C)C)OC(=O)C2=CC=CC=C2C(=O)OC3CC(CC(C3)(C)C)C

Specifications

Melting Point 93°C
Color White
Quantity 5 g
Formula Weight 414.59
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(cis-3,3,5-trimethylcyclohexyl) Phthalate
EINECSNumber (3)-4473
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.