accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHHgClNOO

CAS RN 24579-90-6

IUPAC Name: chloro(2-hydroxy-5-nitrophenyl)mercury
Synonyms: chloro(2-hydroxy-5-nitrophenyl)mercury
Molecular Weight (g/mol): 374.14
Molecular Formula: C6H4ClHgNO3
InChi Key: VIMWCINSBRXAQH-UHFFFAOYSA-M
SMILES: OC1=CC=C(C=C1[Hg]Cl)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)