Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloromercuri-4-nitrophenol, Thermo Scientific™

Catalog No. AC404690050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC404690050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC404690050 Supplier Thermo Scientific Chemicals Supplier No. 404690050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24579-90-6
Molecular Formula C6H4ClHgNO3
Molecular Weight (g/mol) 374.144
InChI Key VIMWCINSBRXAQH-UHFFFAOYSA-M
Synonym 2-chloromercuri-4-nitrophenol, mercury, chloro 2-hydroxy-5-nitrophenyl, chloro 2-hydroxy-5-nitrophenyl mercury, chloro-2-hydroxy-5-nitrophenyl mercury, mercury,chloro 2-hydroxy-5-nitrophenyl
PubChem CID 90545
IUPAC Name chloro-(2-hydroxy-5-nitrophenyl)mercury
SMILES C1=CC(=C(C=C1[N+](=O)[O-])[Hg]Cl)O

Specifications

Color Yellow
Infrared Spectrum Authentic
Quantity 5 g
Formula Weight 374.14
Percent Purity ≥52.5% (Hg)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloromercuri-4-nitrophenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.