accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(S)NONHONOHNO

CAS RN 24584-09-6

IUPAC Name: 4-[(2S)-2-(3,5-dioxopiperazin-1-yl)propyl]piperazine-2,6-dione
Synonyms: dexrazoxane dexrazoxano [inn-spanish] (+)-dexrazoxane dexrazoxano dextrorazoxane razoxana [inn-spanish] dexrazoxanum razoxanum desrazoxane
Molecular Weight (g/mol): 268.27
Molecular Formula: C11H16N4O4
InChi Key: BMKDZUISNHGIBY-ZETCQYMHSA-N
SMILES: C[C@@H](CN1CC(=O)NC(=O)C1)N1CC(=O)NC(=O)C1
Percent Purity 
  • (1)
  • (1)
Grade 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)