Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dexrazoxane 98.0+%, TCI America™
SDP

Catalog No. D4227100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4227100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4227100MG Supplier TCI America Supplier No. D4227100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 24584-09-6
Molecular Formula C11H16N4O4
Molecular Weight (g/mol) 268.273
MDL Number MFCD00866449
InChI Key BMKDZUISNHGIBY-ZETCQYMHSA-N
Synonym (S)-4,4′C-(Propane-1,2-diyl)bis(piperazine-2,6-dione), (S)-1,2-Bis(3,5-dioxo-1-piperazinyl)propane
PubChem CID 71384
ChEBI CHEBI:50223
IUPAC Name 4-[(2S)-2-(3,5-dioxopiperazin-1-yl)propyl]piperazine-2,6-dione
SMILES CC(CN1CC(=O)NC(=O)C1)N2CC(=O)NC(=O)C2

Specifications

Melting Point 195°C
Quantity 100 mg
Formula Weight 268.27
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dexrazoxane
RTECSNumber TL6390000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.