accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOH

CAS RN 2467-03-0

IUPAC Name: 4-[(2-hydroxyphenyl)methyl]phenol
Synonyms: 4-[(2-hydroxyphenyl)methyl]phenol
Molecular Weight (g/mol): 200.24
Molecular Formula: C13H12O2
InChi Key: LVLNPXCISNPHLE-UHFFFAOYSA-N
SMILES: OC1=CC=C(CC2=CC=CC=C2O)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results