accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2469-45-6

IUPAC Name: 1-(dodecylsulfanyl)dodecane
Synonyms: 1-(dodecylsulfanyl)dodecane
Molecular Weight (g/mol): 370.72
Molecular Formula: C24H50S
InChi Key: UPYPTOCXMIWHSG-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCSCCCCCCCCCCCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results