Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dodecyl Sulfide 96.0+%, TCI America™
SDP

Catalog No. D1327500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1327500G 500 g
D132725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1327500G Supplier TCI America Supplier No. D1327500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2469-45-6
Molecular Formula C24H50S
Molecular Weight (g/mol) 370.724
MDL Number MFCD00008975
InChI Key UPYPTOCXMIWHSG-UHFFFAOYSA-N
Synonym dodecyl sulfide, didodecyl sulfide, dilauryl sulfide, n-dodecyl sulfide, dodecane, 1,1'-thiobis, bis dodecyl sulfide, bis n-dodecyl sulfide, unii-m8cf2nuw66, m8cf2nuw66, dodecylthiododecane
PubChem CID 72875
IUPAC Name 1-dodecylsulfanyldodecane
SMILES CCCCCCCCCCCCSCCCCCCCCCCCC

Specifications

Melting Point 41°C
Color White-Yellow
Quantity 500 g
Formula Weight 370.72
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dodecyl Sulfide
EINECSNumber (2)-0473&(2)-0475
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.