accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3(R)(R)H3CCH3OH3CO

CAS RN 24691-16-5

IUPAC Name: (1R,5R)-3,3,5-trimethylcyclohexyl acetate
Synonyms: (1R,5R)-3,3,5-trimethylcyclohexyl acetate
Molecular Weight (g/mol): 184.28
Molecular Formula: C11H20O2
InChi Key: OIVWFAFCHQDCCG-WCBMZHEXSA-N
SMILES: C[C@H]1C[C@H](CC(C)(C)C1)OC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results