Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-3,3,5-Trimethylcyclohexyl Acetate 97.0+%, TCI America™
SDP

Catalog No. A122425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A122425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A122425ML Supplier TCI America Supplier No. A122425ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 24691-16-5
Molecular Formula C11H20O2
Molecular Weight (g/mol) 184.279
MDL Number MFCD00036661
InChI Key OIVWFAFCHQDCCG-WCBMZHEXSA-N
Synonym Acetic Acid cis-3,3,5-Trimethylcyclohexyl Ester
PubChem CID 229657
IUPAC Name [(1R,5R)-3,3,5-trimethylcyclohexyl] acetate
SMILES CC1CC(CC(C1)(C)C)OC(=O)C

Specifications

Color Yellow
Boiling Point 97°C
Quantity 25 mL
Formula Weight 184.28
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-3,3,5-Trimethylcyclohexyl Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.